siRNA Details
Gene Symbol: IZUMO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031735.2 | NP_001026905.2 | 1 | 246 | ||
NM_001039846.1 | NP_001034935.1 | 1 | 246 | |||
XM_005259480.3 | XP_005259537.2 | 1 | 267 | |||
GenBank mRNA | AK294083.1 | - | 1 | 117 | ||
AK301957.1 | - | 1 | 200 | |||
AY358444.1 | - | 1 | 246 | |||
AY753639.1 | - | 1 | 205 | |||
BC010443.1 | - | 1 | 185 | |||
BC014609.2 | - | 1 | 200 | 4825119 | ||
BC040964.1 | - | 1 | 187 | |||
BI827974.1 | - | 1 | 103 | |||
BX378854.2 | - | 1 | 246 | |||
DQ196309.1 | - | 1 | 233 | |||
DQ196310.1 | - | 1 | 233 | |||
DQ424900.1 | - | 1 | 169 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
