siRNA Details
Gene Symbol: PPP1R14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243947.1 | NP_001230876.1 | 1 | 413 | ||
NM_033256.2 | NP_150281.1 | 1 | 413 | |||
GenBank mRNA | AB056508.1 | - | 1 | 354 | ||
AB056509.1 | - | 1 | 354 | |||
AF123763.1 | - | 1 | 204 | |||
AY050670.1 | - | 1 | 198 | |||
BC021089.1 | - | 1 | 178 | 5019808 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
