siRNA Details
Gene Symbol: TBX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005995.4 | NP_005986.2 | 4, 5 | 645 | ||
XM_011544992.2 | XP_011543294.1 | 3, 4 | 935 | |||
XM_011544993.1 | XP_011543295.1 | 2, 3 | 271 | |||
GenBank mRNA | AY229977.1 | - | 4, 5 | 646 | ||
BC113485.1 | - | 4, 5 | 639 | 8322537 | ||
BC113487.1 | - | 4, 5 | 639 | 8322539 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
