siRNA Details
Gene Symbol: IRF2BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015649.2 | NP_056464.1 | 1 | 734 | ||
GenBank mRNA | AK223622.1 | - | 1 | 614 | ||
AK315060.1 | - | 1 | 674 | |||
AY278022.1 | - | 1 | 685 | |||
BC005834.2 | - | 1 | 122 | |||
BC038222.1 | - | 1 | 682 | 5168442 | ||
BC039002.1 | - | 1 | 699 | 5182566 | ||
BC078181.1 | - | 1 | 407 | 6501537 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
