siRNA Details
Gene Symbol: PODXL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015720.3 | NP_056535.1 | 3 | 684 | ||
GenBank mRNA | AF219137.1 | - | 3 | 643 | ||
AL524055.3 | - | 3 | 683 | |||
AY359096.1 | - | 3 | 647 | |||
BC019330.1 | - | 3 | 659 | 3502164 | ||
BC052585.1 | - | 3 | 639 | 6264322 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
