siRNA Details
Gene Symbol: ZBTB46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025224.3 | NP_079500.2 | 5 | 4351 | ||
XM_005260195.4 | XP_005260252.1 | 5 | 4431 | |||
XM_005260196.3 | XP_005260253.1 | 5 | 4247 | |||
XM_005260197.4 | XP_005260254.1 | 5 | 4567 | |||
XM_005260198.4 | XP_005260255.1 | 5 | 4230 | |||
XM_006723700.3 | XP_006723763.1 | 5 | 4263 | |||
XM_011528548.2 | XP_011526850.1 | 5 | 4531 | |||
GenBank mRNA | AK023564.1 | - | 1 | 1335 | ||
AK126720.1 | - | 4 | 3368 | |||
AK131482.1 | - | 5 | 4351 | |||
BC052269.1 | - | 7 | 2311 | 6574149 | ||
BC073800.1 | - | 7 | 2311 | 6576441 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
