siRNA Details
Gene Symbol: ENHO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198573.2 | NP_940975.2 | 2 | 810 | ||
GenBank mRNA | AY358658.1 | - | 1 | 538 | ||
BC022101.1 | - | 3 | 619 | 4871461 | ||
BQ932712.1 | - | 2 | 661 | |||
BX111579.1 | - | 1 | 397 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
