siRNA Details
Gene Symbol: UTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003577.2 | NP_003568.2 | 1 | 308 | ||
GenBank mRNA | AB011076.2 | - | 1 | 309 | ||
AY606112.1 | - | 1 | 308 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
