siRNA Details
Gene Symbol: HIST3H3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003493.2 | NP_003484.1 | 1 | 18 | ||
GenBank mRNA | AK312217.1 | - | 1 | 51 | ||
BC069079.1 | - | 1 | 18 | 7216893 | ||
BC101837.1 | - | 1 | 66 | 8069343 | ||
BC101839.1 | - | 1 | 66 | 8069345 | ||
CR542013.1 | - | 1 | 18 | |||
DQ587912.1 | - | 1 | 17 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
