siRNA Details
Gene Symbol: SCAND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016558.3 | NP_057642.1 | 3 | 1241 | ||
NM_033630.2 | NP_361012.2 | 2 | 772 | |||
GenBank mRNA | AF204271.1 | - | 1 | 737 | ||
AF207829.1 | - | 2 | 652 | |||
BC000785.2 | - | 2 | 636 | 3508136 | ||
BC036709.2 | - | 3 | 1609 | 4620185 | ||
BC041022.1 | - | 3 | 1609 | 3830122 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
