siRNA Details
Gene Symbol: IP6K2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005909.2 | NP_001005909.1 | 2, 3 | 459 | ||
NM_016291.3 | NP_057375.2 | 2, 3 | 462 | |||
XM_006713199.3 | XP_006713262.1 | 3, 4 | 566 | |||
XM_006713200.1 | XP_006713263.1 | 3, 4 | 536 | |||
XM_006713201.1 | XP_006713264.1 | 3, 4 | 613 | |||
XM_006713202.1 | XP_006713265.1 | 3, 4 | 615 | |||
XM_011533816.1 | XP_011532118.1 | 3, 4 | 596 | |||
XM_017006583.1 | XP_016862072.1 | 2, 3 | 458 | |||
XM_017006584.1 | XP_016862073.1 | 2, 3 | 433 | |||
XM_017006585.1 | XP_016862074.1 | 3, 4 | 548 | |||
GenBank mRNA | AF177145.1 | - | 1, 2 | 356 | ||
AK024045.1 | - | 2, 3 | 399 | |||
AK315204.1 | - | 2, 3 | 400 | |||
AY007091.1 | - | 2, 3 | 385 | |||
BC019694.1 | - | 2, 3 | 371 | 4941646 | ||
BC065533.1 | - | 2, 3 | 388 | 6162048 | ||
DB039955.1 | - | 2, 3 | 459 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
