siRNA Details
Gene Symbol: TMC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127198.2 | NP_001120670.1 | 8 | 996 | ||
NM_001321185.1 | NP_001308114.1 | 8 | 949 | |||
NM_007267.7 | NP_009198.4 | 8 | 900 | |||
XM_005256995.1 | XP_005257052.1 | 8 | 1025 | |||
XM_011524255.1 | XP_011522557.1 | 7 | 927 | |||
XM_011524258.1 | XP_011522560.1 | 7 | 927 | |||
XM_017024107.1 | XP_016879596.1 | 8 | 974 | |||
XM_017024108.1 | XP_016879597.1 | 8 | 927 | |||
XM_017024109.1 | XP_016879598.1 | 6 | 716 | |||
GenBank mRNA | AK021738.1 | - | 6 | 757 | ||
AK074065.1 | - | 8 | 899 | |||
AK096076.1 | - | 8 | 932 | |||
AK303165.1 | - | 4 | 473 | |||
AK310734.1 | - | 7 | 1320 | |||
AY057379.1 | - | 7 | 739 | |||
AY099356.1 | - | 9 | 898 | |||
AY099357.1 | - | 9 | 898 | |||
AY236496.1 | - | 7 | 865 | |||
BC023597.2 | - | 8 | 880 | 4566166 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
