siRNA Details
Gene Symbol: TMC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127198.2 | NP_001120670.1 | 8 | 1074 | ||
NM_001321185.1 | NP_001308114.1 | 8 | 1027 | |||
NM_007267.7 | NP_009198.4 | 8 | 978 | |||
XM_005256995.1 | XP_005257052.1 | 8 | 1103 | |||
XM_011524255.1 | XP_011522557.1 | 7 | 1005 | |||
XM_011524256.1 | XP_011522558.1 | 7 | 879 | |||
XM_011524257.2 | XP_011522559.1 | 2 | 123 | |||
XM_011524258.1 | XP_011522560.1 | 7 | 1005 | |||
XM_017024107.1 | XP_016879596.1 | 8 | 1052 | |||
XM_017024108.1 | XP_016879597.1 | 8 | 1005 | |||
XM_017024109.1 | XP_016879598.1 | 6 | 794 | |||
GenBank mRNA | AK021738.1 | - | 6 | 835 | ||
AK074065.1 | - | 8 | 977 | |||
AK096076.1 | - | 8 | 1010 | |||
AK303165.1 | - | 4 | 551 | |||
AK310734.1 | - | 7 | 1398 | |||
AY057379.1 | - | 7 | 817 | |||
AY099356.1 | - | 9 | 976 | |||
AY099357.1 | - | 9 | 976 | |||
AY236496.1 | - | 7 | 943 | |||
BC007766.1 | - | 7 | 717 | |||
BC023597.2 | - | 8 | 958 | 4566166 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
