siRNA Details
Gene Symbol: TMC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127198.2 | NP_001120670.1 | 10 | 1465 | ||
NM_001321185.1 | NP_001308114.1 | 10 | 1418 | |||
NM_007267.7 | NP_009198.4 | 10 | 1369 | |||
XM_005256995.1 | XP_005257052.1 | 10 | 1494 | |||
XM_011524255.1 | XP_011522557.1 | 9 | 1396 | |||
XM_011524256.1 | XP_011522558.1 | 9 | 1270 | |||
XM_011524257.2 | XP_011522559.1 | 4 | 514 | |||
XM_011524258.1 | XP_011522560.1 | 9 | 1396 | |||
XM_017024107.1 | XP_016879596.1 | 10 | 1443 | |||
XM_017024108.1 | XP_016879597.1 | 10 | 1396 | |||
XM_017024109.1 | XP_016879598.1 | 8 | 1185 | |||
GenBank mRNA | AB002405.2 | - | 2 | 177 | ||
AK021738.1 | - | 8 | 1226 | |||
AK074065.1 | - | 10 | 1368 | |||
AK096076.1 | - | 10 | 1401 | |||
AK310734.1 | - | 9 | 1789 | |||
AY057379.1 | - | 9 | 1208 | |||
AY099356.1 | - | 11 | 1367 | |||
AY099357.1 | - | 11 | 1367 | |||
AY236496.1 | - | 9 | 1334 | |||
BC007766.1 | - | 9 | 1108 | |||
BC018346.1 | - | 3 | 333 | |||
BC023597.2 | - | 10 | 1349 | 4566166 | ||
BC035648.1 | - | 3 | 389 | 5575949 | ||
DQ104440.1 | - | 1 | 125 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
