siRNA Details
Gene Symbol: TLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015059.2 | NP_055874.2 | 6 | 748 | ||
XM_005254708.4 | XP_005254765.1 | 9 | 1116 | |||
XM_005254710.4 | XP_005254767.1 | 10 | 1254 | |||
XM_005254711.4 | XP_005254768.1 | 9 | 1476 | |||
XM_005254712.4 | XP_005254769.1 | 9 | 1133 | |||
XM_005254713.4 | XP_005254770.1 | 9 | 2030 | |||
XM_005254714.3 | XP_005254771.1 | 8 | 1019 | |||
XM_005254715.2 | XP_005254772.1 | 8 | 1725 | |||
XM_006720717.3 | XP_006720780.1 | 9 | 1850 | |||
XM_017022665.1 | XP_016878154.1 | 9 | 2201 | |||
XM_017022666.1 | XP_016878155.1 | 9 | 1060 | |||
XM_017022667.1 | XP_016878156.1 | 7 | 922 | |||
XM_017022668.1 | XP_016878157.1 | 9 | 1849 | |||
XM_017022669.1 | XP_016878158.1 | 9 | 1850 | |||
GenBank mRNA | AF402000.1 | - | 6 | 748 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
