siRNA Details
Gene Symbol: TLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015059.2 | NP_055874.2 | 11 | 1259 | ||
XM_005254708.4 | XP_005254765.1 | 14 | 1627 | |||
XM_005254710.4 | XP_005254767.1 | 15 | 1765 | |||
XM_005254711.4 | XP_005254768.1 | 14 | 1987 | |||
XM_005254712.4 | XP_005254769.1 | 14 | 1644 | |||
XM_005254713.4 | XP_005254770.1 | 14 | 2541 | |||
XM_005254714.3 | XP_005254771.1 | 13 | 1530 | |||
XM_005254715.2 | XP_005254772.1 | 13 | 2236 | |||
XM_006720717.3 | XP_006720780.1 | 14 | 2361 | |||
XM_017022665.1 | XP_016878154.1 | 14 | 2712 | |||
XM_017022666.1 | XP_016878155.1 | 14 | 1571 | |||
XM_017022667.1 | XP_016878156.1 | 12 | 1433 | |||
XM_017022668.1 | XP_016878157.1 | 14 | 2360 | |||
XM_017022669.1 | XP_016878158.1 | 14 | 2361 | |||
GenBank mRNA | AF402000.1 | - | 11 | 1259 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
