siRNA Details
Gene Symbol: TLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015059.2 | NP_055874.2 | 47, 48 | 6476 | ||
XM_005254708.4 | XP_005254765.1 | 50, 51 | 6844 | |||
XM_005254710.4 | XP_005254767.1 | 51, 52 | 6982 | |||
XM_005254711.4 | XP_005254768.1 | 50, 51 | 7204 | |||
XM_005254712.4 | XP_005254769.1 | 50, 51 | 6861 | |||
XM_005254713.4 | XP_005254770.1 | 50, 51 | 7758 | |||
XM_005254714.3 | XP_005254771.1 | 49, 50 | 6747 | |||
XM_005254715.2 | XP_005254772.1 | 49, 50 | 7453 | |||
XM_006720717.3 | XP_006720780.1 | 50, 51 | 7578 | |||
XM_017022665.1 | XP_016878154.1 | 50, 51 | 7929 | |||
XM_017022666.1 | XP_016878155.1 | 50, 51 | 6788 | |||
XM_017022667.1 | XP_016878156.1 | 48, 49 | 6650 | |||
XM_017022668.1 | XP_016878157.1 | 50, 51 | 7577 | |||
XM_017022669.1 | XP_016878158.1 | 49, 50 | 7443 | |||
GenBank mRNA | AB002318.2 | - | 33, 34 | 4650 | ||
AB621808.1 | - | 28, 29 | 3759 | |||
AF402000.1 | - | 47, 48 | 6476 | |||
BX647654.1 | - | 18, 19 | 4390 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
