siRNA Details
Gene Symbol: IGF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000618.4 | NP_000609.1 | 3 | 502 | ||
NM_001111283.2 | NP_001104753.1 | 3 | 502 | |||
NM_001111284.1 | NP_001104754.1 | 3 | 340 | |||
XM_017019259.1 | XP_016874748.1 | 3 | 312 | |||
XM_017019260.1 | XP_016874749.1 | 3 | 531 | |||
XM_017019261.1 | XP_016874750.1 | 3 | 1991 | |||
XM_017019262.1 | XP_016874751.1 | 3 | 312 | |||
XM_017019263.1 | XP_016874752.1 | 3 | 304 | |||
GenBank mRNA | AB209184.1 | - | 3 | 453 | ||
AK312231.1 | - | 3 | 401 | |||
AL710591.1 | - | 3 | 503 | |||
BC148266.1 | - | 3 | 260 | 8860206 | ||
BC152321.1 | - | 3 | 383 | 40129266 | ||
CR541861.1 | - | 3 | 238 | |||
M11568.1 | - | 3 | 420 | |||
M27544.1 | - | 3 | 386 | |||
M29644.1 | - | 3 | 249 | |||
M37484.1 | - | 3 | 340 | |||
U40870.1 | - | 2 | 181 | |||
X00173.1 | - | 3 | 249 | |||
X56773.1 | - | 3 | 238 | |||
X56774.1 | - | 3 | 238 | |||
X57025.1 | - | 3 | 404 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
