siRNA Details
Gene Symbol: C2orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085437.2 | NP_001078906.2 | 1, 2 | 678 | ||
XM_011511877.1 | XP_011510179.1 | 2, 3 | 670 | |||
GenBank mRNA | AK055735.1 | - | 1, 2 | 1361 | ||
AK056601.1 | - | 1, 2 | 678 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
