siRNA Details
Gene Symbol: C2orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085437.2 | NP_001078906.2 | 3 | 1031 | ||
NM_001282921.1 | NP_001269850.1 | 3 | 524 | |||
NM_024861.3 | NP_079137.3 | 3 | 467 | |||
XM_011511877.1 | XP_011510179.1 | 4 | 1023 | |||
GenBank mRNA | AK026324.1 | - | 3 | 464 | ||
AK055735.1 | - | 3 | 1714 | |||
AK056601.1 | - | 3 | 1031 | |||
AK096434.1 | - | 3 | 524 | |||
BC125154.1 | - | 3 | 455 | 40120634 | ||
BC125155.1 | - | 3 | 455 | 40120635 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
