siRNA Details
Gene Symbol: RNASEH2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032193.3 | NP_115569.2 | 4 | 713 | ||
GenBank mRNA | AF346606.1 | - | 4 | 713 | ||
AK024627.1 | - | 4 | 570 | |||
AK225131.1 | - | 4 | 570 | |||
AK297660.1 | - | 4 | 483 | |||
BC023588.2 | - | 4 | 593 | 4763204 | ||
BM921525.1 | - | 4 | 582 | |||
L08647.1 | - | 3 | 442 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
