siRNA Details
Gene Symbol: CEP290 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025114.3 | NP_079390.3 | 27 | 3385 | ||
XM_011538756.2 | XP_011537058.1 | 29 | 4246 | |||
XM_011538757.2 | XP_011537059.1 | 29 | 4054 | |||
XM_011538758.2 | XP_011537060.1 | 29 | 4246 | |||
XM_011538759.2 | XP_011537061.1 | 29 | 4246 | |||
XM_011538760.1 | XP_011537062.1 | 29 | 4246 | |||
XM_011538761.1 | XP_011537063.1 | 29 | 4246 | |||
XM_011538762.2 | XP_011537064.1 | 28 | 3478 | |||
XM_011538763.2 | XP_011537065.1 | 27 | 3385 | |||
XM_011538764.2 | XP_011537066.1 | 29 | 4246 | |||
XM_011538765.2 | XP_011537067.1 | 29 | 4246 | |||
XM_011538766.2 | XP_011537068.1 | 15 | 2473 | |||
XM_017019980.1 | XP_016875469.1 | 29 | 4246 | |||
XM_017019981.1 | XP_016875470.1 | 29 | 4246 | |||
XM_017019982.1 | XP_016875471.1 | 29 | 4246 | |||
XM_017019983.1 | XP_016875472.1 | 26 | 3364 | |||
GenBank mRNA | AB002371.1 | - | 2 | 1401 | ||
AK025632.1 | - | 15 | 1988 | |||
AY590151.1 | - | 19 | 2306 | |||
BC008641.1 | - | 3 | 437 | |||
DQ109808.1 | - | 27 | 3385 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
