siRNA Details
Gene Symbol: PHLDA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012396.4 | NP_036528.1 | 1 | 549 | ||
NR_073080.1 | 1 | 549 | ||||
GenBank mRNA | AF151100.1 | - | 1 | 483 | ||
AK075179.1 | - | 1 | 512 | |||
AK222644.1 | - | 1 | 512 | |||
AK312115.1 | - | 1 | 180 | |||
BC014390.1 | - | 1 | 401 | 3547186 | ||
BC068273.1 | - | 1 | 549 | 5177273 |

More Information
Other Species Targeted

Gene Ontology Categories:
