siRNA Details
Gene Symbol: DCAKD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128631.2 | NP_001122103.1 | 2 | 300 | ||
NM_001288654.2 | NP_001275583.1 | 2 | 436 | |||
NM_001288655.1 | NP_001275584.1 | 2 | 348 | |||
NM_001321326.1 | NP_001308255.1 | 2 | 246 | |||
NM_024819.6 | NP_079095.3 | 2 | 433 | |||
XM_005257688.2 | XP_005257745.1 | 2 | 295 | |||
XM_011525262.2 | XP_011523564.1 | 2 | 465 | |||
XM_017025102.1 | XP_016880591.1 | 2 | 477 | |||
XM_017025103.1 | XP_016880592.1 | 2 | 477 | |||
XM_017025104.1 | XP_016880593.1 | 2 | 477 | |||
XM_017025105.1 | XP_016880594.1 | 2 | 430 | |||
XM_017025106.1 | XP_016880595.1 | 2 | 372 | |||
GenBank mRNA | AK026608.1 | - | 2 | 300 | ||
AK290755.1 | - | 2 | 249 | |||
BC006472.1 | - | 2 | 322 | 2822306 | ||
BC006546.2 | - | 2 | 322 | 2822306 | ||
BC018132.2 | - | 1 | 194 | 3903964 | ||
BC033299.1 | - | 1 | 333 | 4153431 | ||
BF115235.1 | - | 2 | 309 | |||
BM910634.1 | - | 2 | 348 | |||
BP872609.1 | - | 2 | 412 | |||
BX487139.1 | - | 2 | 410 | |||
DR005747.1 | - | 2 | 348 | |||
HY061203.1 | - | 2 | 246 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
