siRNA Details
Gene Symbol: ZNF621 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098414.2 | NP_001091884.1 | 3 | 293 | ||
NM_001287245.1 | NP_001274174.1 | 3 | 529 | |||
NM_198484.4 | NP_940886.1 | 3 | 529 | |||
GenBank mRNA | AK074366.1 | - | 3 | 454 | ||
AK127181.1 | - | 3 | 293 | |||
BC101619.1 | - | 3 | 167 | 8069125 | ||
BC113415.1 | - | 3 | 167 | 8322467 | ||
CR749268.1 | - | 3 | 394 | |||
DA282973.1 | - | 3 | 293 | |||
DB480668.2 | - | 3 | 519 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
