siRNA Details
Gene Symbol: KLF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138693.3 | NP_619638.2 | 1 | 843 | ||
GenBank mRNA | AF490374.1 | - | 1 | 843 | ||
DQ534757.1 | - | 1 | 917 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
