siRNA Details
Gene Symbol: KCNE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012282.2 | NP_036414.1 | 1 | 355 | ||
GenBank mRNA | AJ012743.1 | - | 1 | 355 | ||
AK223306.1 | - | 1 | 359 | |||
AK314923.1 | - | 1 | 359 | |||
BC035330.1 | - | 1 | 334 | 5197378 | ||
DQ192294.1 | - | 1 | 359 | |||
EF535525.1 | - | 1 | 266 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
