siRNA Details
Gene Symbol: CEBPD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005195.3 | NP_005186.2 | 1 | 207 | ||
GenBank mRNA | BC094715.1 | - | 1 | 459 | ||
BC105109.1 | - | 1 | 202 | 8144112 | ||
M83667.1 | - | 1 | 251 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
