siRNA Details
Gene Symbol: WFDC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080827.1 | NP_543017.1 | 1 | 103 | ||
GenBank mRNA | AF411861.1 | - | 1 | 103 | ||
BC101600.1 | - | 1 | 98 | 8069106 | ||
BC101604.1 | - | 1 | 98 | 8069110 | ||
GQ472217.1 | - | 1 | 98 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
