siRNA Details
Gene Symbol: CALML5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017422.4 | NP_059118.2 | 1 | 305 | ||
GenBank mRNA | AF172852.1 | - | 1 | 286 | ||
AK222744.1 | - | 1 | 305 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
