siRNA Details
Gene Symbol: ARPP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025068.1 | NP_001020239.1 | 6 | 661 | ||
NM_001025069.1 | NP_001020240.1 | 6 | 670 | |||
NM_001267616.1 | NP_001254545.1 | 6 | 1817 | |||
NM_001267618.1 | NP_001254547.1 | 5 | 758 | |||
NM_198399.1 | NP_938409.1 | 5 | 652 | |||
GenBank mRNA | AK055064.1 | - | 6 | 670 | ||
AK289649.1 | - | 5 | 607 | |||
AK289868.1 | - | 6 | 664 | |||
BC017805.1 | - | 5 | 652 | |||
BC066651.1 | - | 3 | 1376 | |||
BX537647.1 | - | 1 | 979 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
