siRNA Details
Gene Symbol: RBAK-RBAKDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204513.1 | NP_001191442.1 | 5, 6 | 551 | ||
GenBank mRNA | BM929719.1 | - | 5, 6 | 461 |
Gene Symbol: RBAKDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015343.2 | 2, 3 | 272 | |||
GenBank mRNA | CR745855.1 | - | 2, 3 | 165 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
