siRNA Details
Gene Symbol: TFAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270782.1 | NP_001257711.1 | 3, 4 | 706 | ||
NM_003201.2 | NP_003192.1 | 3, 4 | 706 | |||
NR_073073.1 | 4, 5 | 773 | ||||
XM_011540120.2 | XP_011538422.1 | 3, 4 | 439 | |||
XM_011540121.2 | XP_011538423.1 | 3, 4 | 439 | |||
GenBank mRNA | AB451241.1 | - | 3, 4 | 301 | ||
AB451366.1 | - | 3, 4 | 301 | |||
AK312558.1 | - | 3, 4 | 444 | |||
BC126366.1 | - | 3, 4 | 389 | 8992082 | ||
BM455062.1 | - | 3, 4 | 415 | |||
BT019658.1 | - | 3, 4 | 301 | |||
BT019659.1 | - | 3, 4 | 301 | |||
CR407653.1 | - | 3, 4 | 301 | |||
EU279428.1 | - | 3, 4 | 329 | |||
M62810.1 | - | 3, 4 | 433 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
