siRNA Details
Gene Symbol: TFAMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001288.1 | 1 | 1094 |
Gene Symbol: TFAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270782.1 | NP_001257711.1 | 6 | 1299 | ||
NM_003201.2 | NP_003192.1 | 7 | 1395 | |||
NR_073073.1 | 8 | 1462 | ||||
GenBank mRNA | BC126366.1 | - | 7 | 1078 | 8992082 | |
M62810.1 | - | 7 | 1122 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
