siRNA Details
Gene Symbol: FRAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012083.2 | NP_036215.1 | 1 | 721 | ||
GenBank mRNA | AB045118.1 | - | 1 | 721 | ||
AF062739.1 | - | 1 | 232 | |||
BC020165.1 | - | 1 | 696 | 3629702 | ||
BT009770.1 | - | 1 | 592 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
