siRNA Details
Gene Symbol: PYY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003064.2 | 2 | 461 | |||
GenBank mRNA | AF222904.1 | - | 2 | 210 | ||
BC093811.1 | - | 1 | 143 | |||
BC093813.1 | - | 1 | 143 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
