siRNA Details
Gene Symbol: KLHDC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184854.1 | NP_001171783.1 | 6 | 729 | ||
NM_001184856.1 | NP_001171785.1 | 7 | 807 | |||
NM_017566.3 | NP_060036.2 | 8 | 900 | |||
XM_005255994.3 | XP_005256051.1 | 8 | 905 | |||
XM_005255999.1 | XP_005256056.1 | 7 | 820 | |||
XM_006721201.1 | XP_006721264.1 | 8 | 920 | |||
XM_006721202.1 | XP_006721265.1 | 8 | 920 | |||
XM_006721203.2 | XP_006721266.1 | 8 | 920 | |||
XM_006721204.3 | XP_006721267.1 | 8 | 920 | |||
XM_017023344.1 | XP_016878833.1 | 8 | 905 | |||
XM_017023345.1 | XP_016878834.1 | 8 | 845 | |||
GenBank mRNA | AK023986.1 | - | 8 | 857 | ||
AK097097.1 | - | 6 | 1781 | |||
AK098630.1 | - | 4 | 348 | |||
AL133107.1 | - | 6 | 663 | |||
BC001044.2 | - | 6 | 636 | 3505126 | ||
BC011680.2 | - | 7 | 791 | 4303424 | ||
BC022969.1 | - | 8 | 844 | 5112326 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
