siRNA Details
Gene Symbol: LOC284241 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136643.1 | 2, 3 | 529 | |||
GenBank mRNA | AK098581.1 | - | 2, 3 | 529 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
