siRNA Details
Gene Symbol: ZBED2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024508.4 | NP_078784.2 | 2 | 990 | ||
GenBank mRNA | BC003536.1 | - | 1 | 175 | 3609995 | |
BC071956.1 | - | 1 | 455 | 3928218 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
