siRNA Details
Gene Symbol: FLJ44635 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207422.2 | NP_997305.2 | 2 | 584 | ||
GenBank mRNA | AK126598.1 | - | 2 | 584 | ||
AY572224.1 | - | 2 | 584 | |||
BC137552.1 | - | 2 | 340 | 9021552 | ||
BC137564.1 | - | 2 | 340 | 9021564 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
