siRNA Details
Gene Symbol: RFT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052859.3 | NP_443091.1 | 13 | 3377 | ||
GenBank mRNA | AK098811.1 | - | 1 | 569 | ||
AL713685.1 | - | 1 | 516 | |||
BC101247.2 | - | 1 | 442 | |||
BC101248.2 | - | 1 | 442 | |||
BC101249.2 | - | 1 | 442 | |||
BC101250.1 | - | 1 | 442 | |||
BC113058.1 | - | 1 | 442 | |||
BC113059.1 | - | 1 | 442 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
