siRNA Details
Gene Symbol: POM121L12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182595.3 | NP_872401.3 | 1 | 410 | ||
GenBank mRNA | AL833888.1 | - | 1 | 61 | ||
BI826445.1 | - | 1 | 409 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
