siRNA Details
Gene Symbol: HID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030630.2 | NP_085133.1 | 19 | 3300 | ||
XM_005257226.1 | XP_005257283.1 | 19 | 3295 | |||
GenBank mRNA | AK074401.1 | - | 19 | 3228 | ||
AK075111.1 | - | 9 | 1825 | |||
AK125514.1 | - | 6 | 2121 | |||
BC032219.1 | - | 15 | 2597 | 5200267 | ||
BC035372.1 | - | 19 | 3216 | 5174235 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
