siRNA Details
Gene Symbol: GPR157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024980.4 | NP_079256.4 | 1 | 501 | ||
XM_005263496.4 | XP_005263553.1 | 1 | 450 | |||
XM_005263497.4 | XP_005263554.1 | 1 | 450 | |||
GenBank mRNA | AK022194.1 | - | 1 | 433 | ||
AK292888.1 | - | 1 | 455 | |||
AK304154.1 | - | 1 | 455 | |||
AY768808.1 | - | 1 | 501 | |||
BC094733.1 | - | 3 | 348 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
