siRNA Details
Gene Symbol: ZDHHC22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174976.2 | NP_777636.2 | 3 | 960 | ||
XM_006720119.3 | XP_006720182.1 | 3 | 1190 | |||
XM_011536661.2 | XP_011534963.1 | 3 | 4829 | |||
GenBank mRNA | AK095612.1 | - | 3 | 676 | ||
AK294840.1 | - | 3 | 959 | |||
BC035881.1 | - | 2 | 339 | |||
BC117676.2 | - | 2 | 785 | 40069732 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
