siRNA Details
Gene Symbol: ZNF273 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021148.2 | NP_066971.2 | 4 | 722 | ||
NR_003099.1 | 5 | 811 | ||||
XM_011515746.2 | XP_011514048.1 | 3 | 657 | |||
XM_017011688.1 | XP_016867177.1 | 4 | 748 | |||
XM_017011689.1 | XP_016867178.1 | 9 | 6736 | |||
XM_017011690.1 | XP_016867179.1 | 8 | 6647 | |||
XM_017011691.1 | XP_016867180.1 | 9 | 3938 | |||
XM_017011692.1 | XP_016867181.1 | 7 | 6122 | |||
XM_017011693.1 | XP_016867182.1 | 8 | 1828 | |||
XM_017011694.1 | XP_016867183.1 | 8 | 1186 | |||
XM_017011695.1 | XP_016867184.1 | 5 | 797 | |||
XM_017011696.1 | XP_016867185.1 | 6 | 1023 | |||
XM_017011697.1 | XP_016867186.1 | 7 | 6558 | |||
XM_017011698.1 | XP_016867187.1 | 6 | 1675 | |||
GenBank mRNA | AK090648.1 | - | 8 | 1829 | ||
AL832810.1 | - | 1 | 385 | |||
BC063818.1 | - | 5 | 777 | 6503687 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
