siRNA Details
Gene Symbol: ZNF273 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021148.2 | NP_066971.2 | 4 | 1478 | ||
NR_003099.1 | 5 | 1567 | ||||
XM_011515746.2 | XP_011514048.1 | 3 | 1413 | |||
XM_017011688.1 | XP_016867177.1 | 4 | 1504 | |||
XM_017011689.1 | XP_016867178.1 | 9 | 7492 | |||
XM_017011690.1 | XP_016867179.1 | 8 | 7403 | |||
XM_017011691.1 | XP_016867180.1 | 9 | 4694 | |||
XM_017011692.1 | XP_016867181.1 | 7 | 6878 | |||
XM_017011693.1 | XP_016867182.1 | 8 | 2584 | |||
XM_017011694.1 | XP_016867183.1 | 8 | 1942 | |||
XM_017011695.1 | XP_016867184.1 | 5 | 1553 | |||
XM_017011696.1 | XP_016867185.1 | 6 | 1779 | |||
XM_017011697.1 | XP_016867186.1 | 7 | 7314 | |||
XM_017011698.1 | XP_016867187.1 | 6 | 2431 | |||
GenBank mRNA | AK090648.1 | - | 8 | 2585 | ||
AL832810.1 | - | 1 | 1141 | |||
BC063818.1 | - | 5 | 1533 | 6503687 | ||
X78932.1 | - | 1 | 438 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
