siRNA Details
Gene Symbol: C10orf53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042427.1 | NP_001035892.1 | 1 | 110 | ||
NM_182554.2 | NP_872360.2 | 1 | 110 | |||
GenBank mRNA | AK091773.1 | - | 1 | 97 | ||
BC028127.1 | - | 1 | 62 | 5164374 | ||
BX102324.1 | - | 1 | 110 | |||
DB043416.1 | - | 1 | 83 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
