siRNA Details
Gene Symbol: C10orf53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182554.2 | NP_872360.2 | 3 | 434 | ||
GenBank mRNA | BC028127.1 | - | 3 | 386 | 5164374 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
