siRNA Details
Gene Symbol: FAM127B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078172.1 | NP_001071640.1 | 1 | 831 | ||
GenBank mRNA | AL117556.1 | - | 1 | 770 | ||
BC000393.2 | - | 1 | 804 | 2821721 | ||
BC008096.1 | - | 1 | 589 | |||
BC019300.1 | - | 1 | 804 | 2821721 | ||
CR457123.1 | - | 1 | 147 | |||
CR542228.1 | - | 1 | 147 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
